Skip to Content
Merck
All Photos(1)

Key Documents

17-20000

Sigma-Aldrich

Magna ChIP® G Tissue Kit

obtain repeatable, reliable & site specific tissue biopsies.

Sign Into View Organizational & Contract Pricing

Select a Size

22 ASSAYS
£566.00

£566.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
22 ASSAYS
£566.00

About This Item

UNSPSC Code:
41105331
NACRES:
NA.84

£566.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

General description

Chromatin Immunoprecipitation (ChIP) is an important technique allowing the researcher to analyze in vivo interactions of proteins with genomic DNA. Any chromatin-associated or DNA binding protein can be analyzed with this technique, provided a good antibody to the protein exists. One can measure different proteins localized to a specific region of the genome, or the genome wide distribution of a specific protein. Another powerful application of this technique is to analyze changes in histone modifications that correlate with processes like transcription, mitosis or DNA repair.

Historically, ChIP protocols have been geared towards homogenous populations of cell culture. In contrast, tissue samples are more complex and the cells are heterogeneous thus making ChIP analysis less amenable. The Magna ChIP G Tissue Kit provides the tools necessary to obtain repeatable, reliable, and site specific tissue biopsies. Microdissected functionally related populations of cells within a heterogeneous tissue can thus be analyzed with ease, precision and certainty.

Application

The Magna ChIP G Tissue Kit provides the tools necessary to obtain repeatable, reliable & site specific tissue biopsies.

Packaging

22 ChIP assays from tissue samples.
Two modules containing all necessary reagents to perform 22 individual chromatin immunoprecipitation (ChIP) reactions from tissue samples.

Components

  • Magnetic Protein G beads
  • ChIP Dilution Buffer
  • Low Salt Wash Buffer
  • High Salt Wash Buffer
  • LiCl Wash Buffer
  • TE Buffer
  • Tissue Lysis Buffer
  • 10X Tissue Stabililizing Solution
  • ChIP Elution buffer (w/o Proteinase K)
  • 10X Glycine
  • 10X PBS
  • 1mm Micro-dissection punch
  • Spin Filters
  • Collection Tubes
  • Bind Reagent A
  • Wash Reagent B
  • Elution Reagent C
  • Protease Inhibitor Cocktail II
  • Proteinase K

Storage and Stability

Upon receipt, store components at the temperatures indicated on the labels. Kit components are stable for 6 months from date of shipment when used and stored as directed.

Legal Information

MAGNA CHIP is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

Flash Point(F)

55.4 °F

Flash Point(C)

13 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service