Skip to Content
Merck
All Photos(1)

Key Documents

1.50269

Supelco

Purospher®STAR Si (5 µm) HPLC Columns

L × I.D. 250 mm x 4 mm LiChroCART HPLC cartridge, manuCART holder required for use

Synonym(s):

Purospher® STAR SI (5 µm) LiChroCART 250-4

Sign Into View Organizational & Contract Pricing

Select a Size

1 UNIT
£617.00

£617.00


Please contact Customer Service for Availability


Select a Size

Change View
1 UNIT
£617.00

About This Item

UNSPSC Code:
41115709
NACRES:
SB.52

£617.00


Please contact Customer Service for Availability

Product Name

Purospher® STAR Si (5 µm), L × I.D. 250 mm x 4 mm LiChroCART HPLC cartridge, manuCART holder required for use

material

stainless steel column

Quality Level

Agency

suitable for USP L3

product line

Purospher®

feature

endcapped: no

packaging

1 ea of

parameter

250 bar pressure
65 °C max. temp.

technique(s)

HPLC: suitable
LC/MS: suitable
mass spectrometry (MS): suitable (High)

column L × I.D.

25 cm × 4 mm

surface area

330 m2/g

matrix

fully porous particle

matrix active group

Silica bonding phase

particle size

5 μm

pore size

120 Å pore size

operating pH range

2-7.5

separation technique

hydrophilic interaction (HILIC)
normal phase

storage temp.

no temp limit

General description

Purospher® STAR Si endcapped, 5 µm HPLC columns offers highest separation efficiency for normal-phase chromatography of low molecular weight compounds soluble in organic solvents.Purospher® STAR Si, 5 µm LiChroCART 250-4 is a cartridge column which requires a manuCART cartridge column holder part number 1.51486.0001. The manuCART can be used to hold one cartridge column with or without direct integration of a 4-4 mm guard column. The recommended guard column is Purospher® STAR RP-8 endcapped (5 µm) LiChroCART 4-4 part number 1.50249.0001. LiChroCART cartridge columns mounted with a manuCART cartridge column holder is hand-tight only and have therefore a maximum pressure stability of 250 bar. The column dimension of 250 mm length x 4 mm I.D. is the optimal choice for the HPLC separation of complex samples allowing significant solvent savings and improved sensitivity.Learn more about Purospher® STAR HPLC and UHPLC ColumnsLearn more about the Supelco HPLC Column portfolio and Method development:Brochure: HPLC and UHPLC Column Selection GuideBrochure: A Practical Guide to HPLC - Method Development Guide

Application

Intended to be used for the separation of aflatoxins, carotenoids, oils, fat-soluble vitamins, lipids, and fatty acids.

Features and Benefits

  • Ultra-pure silica (99.999%) for excellent peak symmetry
  • High separation efficiency
  • Reproducible results from run-to-run and batch-to-batch
  • Use with Normal-phase or HILIC conditions (needs to be flushed to HILIC mobile phase)
  • Good suitability for LC-MS applications

Analysis Note

Pressure: ≤ 65 bar
Theoretical Plates (N/m) (2-Nitrotoluene): ≥ 65000
Symmetry (2-Nitrotoluene): 0.8 - 1.4
Capacity factor (2-Nitrotoluene): 0.5 - 2.0

Legal Information

LICHROCART is a trademark of Merck KGaA, Darmstadt, Germany
MANUCART is a trademark of Merck KGaA, Darmstadt, Germany
Purospher is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10-13 - German Storage Class 10 to 13


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service