Skip to Content
Merck
All Photos(1)

Key Documents

14-936-M

Sigma-Aldrich

NEK9, active

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
£486.00
250 μG
£3,110.00

£486.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 μG
£486.00
250 μG
£3,110.00

About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.32

£486.00


Please contact Customer Service for Availability

Request a Bulk Order

recombinant

expressed in baculovirus infected Sf21 cells

Quality Level

technique(s)

activity assay: suitable

UniProt accession no.

Gene Information

human ... NEK9(91754)

General description

Pleiotropic regulator of mitotic progression, participating in the control of spindle dynamics and chromosome separation. Phosphorylates different histones, myelin basic protein, beta-casein, and BICD2. Phosphorylates histone H3 on serine and threonine residues and beta-casein on serine residues. Important for G1/S transition and S phase progression. Phosphorylates NEK6 and NEK7 and stimulates their activity by releasing the autoinhibitory functions of Tyr-108 and Tyr-97 respectively.

Packaging

Also available in 10μg size as catalog # 14-936 and in bulk as catalog # 14-936-K.
Also available in 250μg size as catalog # 14-936M and in bulk as catalog # 14-936-K.

Quality

Routinely evaluated by phosphorylation of MBP.

Physical properties

41 kDa

Physical form

Protein in 50mM Tris/HCl pH7.5, 300mM NaCl, 20mM β-glycerophosphate, 0.25mM sodium orthovanadate, 10mM NaF, 0.1mM EGTA, 0.03% Brij-35, 270mM sucrose, 1mM benzamidine, 0.2mM PMSF, 0.1% 2-mercaptoethanol. Frozen solution.

Storage and Stability

6 months at -70°C

Analysis Note

For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service