Skip to Content
Merck
All Photos(1)

Key Documents

14-906-M

Sigma-Aldrich

PI3K-C2α Protein, 250 µg

Sign Into View Organizational & Contract Pricing

Select a Size

250 μG
£3,120.00

£3,120.00


Please contact Customer Service for Availability


Select a Size

Change View
250 μG
£3,120.00

About This Item

UNSPSC Code:
12352204
eCl@ss:
32160405
NACRES:
NA.32

£3,120.00


Please contact Customer Service for Availability

biological source

human

Quality Level

mol wt

Mw 161 kDa

purified by

immobilized metal affinity chromatography (IMAC)

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable (kinase)

NCBI accession no.

UniProt accession no.

General description

Expressed by baculovirus in Sf21 insect cells
N-terminal 6His-tagged recombinant human PI3K-C2α amino acids 299–end.

Application

N-terminal 6His-tagged recombinant human PI3K-C2α amino acids 299–end. For use in Kinase Assays.
Research Category
Signaling
Research Sub Category
Inflammation & Autoimmune Mechanisms|Oncology|Diabetes

Biochem/physiol Actions

  • Protein Target: PI3K-C2α
  • Target Sub-Family:PI3K/PI4K

Packaging

Also available in
10 μg
size (cat#14-906) and in
bulk
(cat# 14-906-K).

Quality

Routinely evaluated in the PIProfiler.

Physical form

10 μg of enzyme in 10.6 μl of 50 mM Tris/HCl, pH 7.5, 300 mM NaCl, 0.1 mM EGTA, 0.03% Brij-35, 270 mM sucrose, 0.2 mM PMSF, 1mM benzamidine, 0.1% 2-mercaptoethanol, 20 mM β-glycerophosphate, 0.25 mM Na3VO4, 10 mM NaF.

Storage and Stability

6 months at -70°C

Other Notes

For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service