Skip to Content
Merck
All Photos(1)

Key Documents

1.46680

Millipore

Casein Broth

With Tween® 1 %, Lecithin 5 g, bottle capacity 500 mL, bottle filling volume 500 mL, closure type, Blue screw cap with septum and protector, pack of 6 bottles

Synonym(s):

Casein Lecithin Tween® Broth

Sign Into View Organizational & Contract Pricing

Select a Size

6 EA
£138.00

£138.00


Estimated to ship on22 April 2025Details



Select a Size

Change View
6 EA
£138.00

About This Item

UNSPSC Code:
41106225

£138.00


Estimated to ship on22 April 2025Details


Agency

EP 2.6.1
JP 4.06
USP 71
71

Quality Level

sterility

sterile; autoclaved

form

liquid

feature

closure type Blue screw cap with septum and protector
ready-to-use

composition

Lecithin, 5 g
With Tween®, 1%

packaging

pack of 6 bottles

technique(s)

direct inoculation: suitable
sterile filtration: suitable

bottle capacity

500 mL

bottle filling volume

500 mL

pH

7.1±0.2

application(s)

cosmetics
food and beverages
pharmaceutical

storage temp.

2-25°C

suitability

nonselective for

General description

Casein LT Broth with neutralizers (Tween® 80 and Lecithin) is a complex nutrient rich inhibitor free microbial media designed in compliance with European (EP), Japanese (JP) and United States Pharmacopeia (USP).
The medium is supplemented with peptones from casein and soymeal to provide an enhanced carbon source, which promotes the growth of the microbes present in the test sample.
Lecithin and Polysorbate 80, are effective against quaternary ammonium compounds, amphoteric surfactants, benzamidines, chlorhexidines and dequadin. Polysorbate 80 inactivates benzyl alcohol, carbanilides, dichlorbenzyl alcohols, benzoic acid, p-hydroxybenzoic acid and its esters, phenols, phenylethyl alcohols and PHB esters.

Application

Casein LT Broth (Tween® 80 and Lecithin) is used for the isolation and cultivation of fastidious bacteria, yeast, and molds. Casein LT Broth is suitable for sterility testing of substances, preparations, and products from pharmaceutical and food industry.

Legal Information

TWEEN is a registered trademark of Croda International PLC

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service