Skip to Content
Merck
All Photos(1)

Documents

13-118

Sigma-Aldrich

GST-NFκB p50 subunit (δ434-969)

N-terminal GST-tagged, recombinant human NFkB, p50 subunit (D434-969), expressed in E. coli. Purified using glutathione agarose chromatography.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352204
eCl@ss:
32160405
NACRES:
NA.41

biological source

human

Quality Level

recombinant

expressed in E. coli

form

liquid

manufacturer/tradename

Upstate®

NCBI accession no.

UniProt accession no.

shipped in

wet ice

Gene Information

human ... NFKB1(4790)

General description

N-terminal GST-tagged, recombinant human NFkB, p50 subunit (D434-969), expressed in E. coli. Purified using glutathione agarose chromatography. MW = 74kDa.

Application

DNA Binding Assay

Quality

routinely evaluated in a DNA binding assay

Physical form

50mM Tris-HCl, pH 7.5, 150mM NaCl, 0.1mM EGTA, 0.1% βmercaptoethanol, 0.03% Brij-35, 0.2mM PMSF, 1mM benzamidine and 50% glycerol

Storage and Stability

1 year at -20°C

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service