Skip to Content
Merck
All Photos(1)

Key Documents

121739

Millipore

Adonitol

A cell-permeable pentitol that has cryoprotective effect on rat embryos.

Synonym(s):

Adonitol, Ribitol

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352201
NACRES:
NA.21

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

colorless

solubility

water: 100 mg/mL

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C5H12O5/c6-1-3(8)5(10)4(9)2-7/h3-10H,1-2H2

InChI key

HEBKCHPVOIAQTA-UHFFFAOYSA-N

General description

A cell-permeable pentitol that has a cryoprotective effect on rat embryos.
A cell-permeable pentitol that has cryoprotective effect on rat embryos.

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, refrigerate at (4°C). Stock solutions are stable for up to 2 months at 4°C. Discard if any visible signs of growth appear.

Other Notes

Utsumi, K., et al. 1992. Cryobiology 29, 332.
Yoshioka, S., et al. 1991. Clin. Chem.37, 686.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service