Skip to Content
Merck
All Photos(1)

Key Documents

1.19912

Supelco

LiChrolut® EN (40 - 120 µm) 100 mg (bottom) LiChrolut® RP-18 (40 - 63 µm) 200 mg (top) 6 ml standard PP-tubes

package of 30 tubes

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115711

packaging

package of 30 tubes

technique(s)

solid phase extraction (SPE): suitable

storage temp.

no temp limit

General description

30 extraction tubes per package sorbents separated by frits

Legal Information

LICHROLUT is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service