Skip to Content
Merck
All Photos(1)

Documents

1.15230

Millipore

3-[4-(2-Hydroxyethyl)-1-piperazinyl]-propanesulfonic acid

pKa (20 °C)8.0, ≥99% (acidimetric)

Synonym(s):

3-[4-(2-Hydroxyethyl)-1-piperazinyl]-propanesulfonic acid, HEPPS

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H20N2O4S
CAS Number:
Molecular Weight:
252.33
MDL number:
UNSPSC Code:
12161703
EC Index Number:
240-198-8

Assay

≥99% (acidimetric)

pKa (20 °C)

8.0

storage temp.

2-30°C

InChI

1S/C9H20N2O4S/c12-8-7-11-5-3-10(4-6-11)2-1-9-16(13,14)15/h12H,1-9H2,(H,13,14,15)

InChI key

OWXMKDGYPWMGEB-UHFFFAOYSA-N

General description

buffer substance HEPPS

Analysis Note

Assay (acidimetric): ≥ 99 %
Identity (Raman-spectrum): passes test
UV-Absorption (250 nm, 20 g/l, 1 cm, water): ≤ 0.05
Chloride (Cl): ≤ 0.005 %
Sulphate (SO₄): ≤ 0.005 %
Heavy metals (as Pb): ≤ 0.0005 %
Na (Sodium): ≤ 0.005 %
Loss on Drying (105°C): ≤ 0.5 %
Biological buffer, pKa at 20 °C: 8.0

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service