Skip to Content
Merck
All Photos(1)

Key Documents

1.15213

Sigma-Aldrich

Auto-Hemacolor®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116121

General description

Staining set for automatic blood smear staining with the HEMA-TEK Slide Stainer
The Auto-Hemacolorsystem was developed for the automatic staining of blood and bone marrow smears in conjunction with the HEMA-TEK (Siemens Healthcare Diagnostics) slider stainer. The kit contains a staining solution, a buffer and a washing solution. Staining is achieved by the alkaline dye (azure) binding to the acidic builders of the cells, e.g. chromatin, spongioplasm, and the acidic dye (eosin) binding to the alkaline constituents, e.g. cytoplasm. It is developed and registered as CE / IVD product for applications in diagnostics and can be applied for up to 1000 stains.

Application

Staining set for staining blood smears with the HEMA-TEK Slide Stainer

Legal Information

HEMACOLOR is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service