Skip to Content
Merck
All Photos(1)

Key Documents

1151158

Millipore

ProSep®-vA Ultra 1 x 100 mL

High-titer antibody purification

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23151816
eCl@ss:
32011308

Quality Level

manufacturer/tradename

Prosep®

parameter

<1000 cm/hr flow rate

technique(s)

protein purification: suitable

bed H

≤450 mm

particle size

75-125 μm

pH

1 – 9

Packaging

Bottle

Legal Information

PROSEP is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service