Skip to Content
Merck
All Photos(1)

Documents

1.12117

Millipore

L-Citrulline

≥99% (perchloric acid titration)

Synonym(s):

L-Citrulline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H13N3O3
CAS Number:
Molecular Weight:
175.19
MDL number:
UNSPSC Code:
12352209
EC Index Number:
206-759-6

Assay

≥99% (perchloric acid titration)

form

solid

storage temp.

2-30°C

InChI

1S/C6H13N3O3/c7-4(5(10)11)2-1-3-9-6(8)12/h4H,1-3,7H2,(H,10,11)(H3,8,9,12)

InChI key

RHGKLRLOHDJJDR-UHFFFAOYSA-N

General description

for biochemistry

Analysis Note

Assay (perchloric acid titration): ≥ 99
Identity (IR-spectrum): passes test
Spec. rotation (α 20/D; 2 %; hydrochloric acid 1 mol/l): +22.0 to +23.0
Heavy metals (as Pb): ≤ 0.001 %
NH₄ (Ammonium): ≤ 0.01 %
Foreign amino acids: ≤ 0.3 %
Other ninhydrine positive substances: ≤ 0.1 %

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service