Skip to Content
Merck
All Photos(1)

Key Documents

1.10029

Supelco

Total Hardness Test

colorimetric, water hardness 3-21 °d, water hardness 4-26 °e, water hardness 5-37 °f, MQuant®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116116

description

semi-quantitative

Quality Level

product line

MQuant®

specific analyte(s)

calcium, magnesium

water hardness measuring range

3-21 °d
4-26 °e
5-37 °f

detection method

colorimetric

General description

Method: colorimetric with test strips (single test strips) <3 - >4 - >7 - >14 - >21°d <4 - >5 - >9 - >18 - >26°e <5 - >7 - >12 - >25 - >37°f MQuant®
The total hardness test can be used to measure water hardness. Water hardness is caused by the presence of alkaline earths, primarily calcium and magnesium. Total hardness is measured as the sum of all calcium and magnesium ions.

Legal Information

MQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service