Skip to Content
Merck
All Photos(2)

Documents

1.07212

Millipore

Peptone from soymeal

papain-digested, suitable for microbiology

Synonym(s):

Peptone from soymeal, Fermtech® base materials for biotechnology, Soya Peptone (papainic), Peptone from soymeal

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
41106212
EC Index Number:
293-434-7
NACRES:
NA.74

biological source

soybean

Quality Level

Assay

9.3-10.7% amino-nitrogen basis (purity)

form

medium granules

shelf life

limited shelf life, expiry date on the label

packaging

pkg of 25 kg
pkg of 5 kg
pkg of 500 g

color

light yellow

pH

5.5-7.0 (20 °C, 20 g/L in H2O)

solubility

2200 g/L

bulk density

420 kg/m3

application(s)

microbiology

storage temp.

15-25°C

General description

Peptone from soymeal is a widely used, non-animal peptone used in industrial microbiology as a nutrient source for the growth of microorganisms in large-scale fermentation processes. Peptone from soymeal, also known as soy peptone or peptic soy peptone, is a water-soluble protein hydrolysate obtained from the papainic enzyme digestion of soybean meal and used as a nutrient source in microbial culture media. It is characterized by its high concentration of vitamins and carbohydrates. Its nitrogen content, combined with the naturally occurring vitamins and high carbohydrate content, facilitates rapid and profuse growth of fastidious microorganisms and the resuscitation of sub-lethally injured microorganisms. Its high content of fermentable sugars makes soya peptone unsuitable for fermentation studies, such as in media used for the identification of microorganisms based on sugar fermentation. It is mainly applicable in the cultivation of fastidious and heterotrophic microbes that require complex nutrient sources.

Application

Peptone from soymeal is used in cell culture, molecular genetic media, microbiological assay procedures, and as a nutritive substrate for the cultivation and resuscitation of a large variety of microorganisms. It is commonly used as an ingredient in many microbial culture media in the pharmaceutical, environmental, food and beverage industries.

Features and Benefits

Our Granulated media are:
  • Safer - Significant reduction of inhalation of toxic and allergenic substances resulting in fewer allergic responses. Less contamination of working tools and environment
  • Accurate - No separation of components and no clumping, even under humid or warm conditions, ensure a uniform product with higher performance accuracy
  • Fast - Rapid and uniform dissolution in water
  • Easier - Easier handling and weighing due to the better flow properties. Media do not stick to the flask or container
  • Reliable - Homogenous distribution of ingredients guarantees high reproducibility even if only small amounts of media are used.

Linkage

Replaces: 1.07212.9027

Analysis Note

Appearance: Lightly yellow to lightly brownishyellow granulate.
Identity: passes test
Total nitrogen (N) (Kjeldahl): 9.3 - 10.7 %
Amino nitrogen (as N): 1.8 - 3.2 %
pH-value (5 %; water): 6.5 - 7.5
Sulfated ash (800 °C): ≤ 15.0 %
Loss on Drying (105°C): ≤ 6.0 %
Suitability for microbiology: passes test

Legal Information

FERMTECH is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service