Skip to Content
Merck
All Photos(1)

Documents

1.06687

Supelco

Sodium azide tablets

for the preservation of milk samples for testing one tablet 0.1 g (1 tablet contains about 8 mg sodium azide)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352402
NACRES:
SE.11

Quality Level

pH

8.2 (20 °C, 100 g/L in H2O)

solubility

250 g/L

storage temp.

no temp limit

Analysis Note

Application test: conforms

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2

Supplementary Hazards

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service