Skip to Content
Merck

1.04956

SAFC

tri-Potassium citrate monohydrate

EMPROVE® ESSENTIAL, Ph. Eur., BP, USP, FCC, E 332

Pharma Manufacturing

Synonym(s):

tri-Potassium citrate monohydrate, Tripotassium citrate, Citric acid potassium salt, 2-Hydroxy-1,2,3-Propanetriccarboxylic Acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H5K3O7 · xH2O
CAS Number:
Molecular Weight:
306.39 (anhydrous basis)
MDL number:
UNSPSC Code:
12352107
E Number:
E332
EC Index Number:
212-755-5

Agency

BP
Ph. Eur.
USP

Quality Level

reg. compliance

FCC

product line

EMPROVE® ESSENTIAL

form

solid

pH

7.5-9.0 (20 °C, 50 g/L in H2O)

mp

>180 °C (Elimination of water of crystallisation)

density

1.98 g/cm3 at 20 °C

application(s)

liquid formulation
ophthalmics
pharmaceutical
semi-solid formulation
solid formulation

storage temp.

15-25°C

InChI

1S/C6H8O7.3K.H2O/c7-3(8)1-6(13,5(11)12)2-4(9)10;;;;/h13H,1-2H2,(H,7,8)(H,9,10)(H,11,12);;;;1H2/q;3*+1;/p-3

InChI key

PJAHUDTUZRZBKM-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

General description

Finding the right excipient that matches your needs as well as regulatory demands can be a complicated challenge in formulation. With our application know-how and regulatory expertise, we support you in every step of development, scale-up, and production.
As part of our Emprove® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.

Application

tri-Potassium citrate is typically used as buffering, alkalizing and sequestering agent. Additional uses include the application as potassium source and as therapeutic ingredient to treat cystitis by alkalizing the urine.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service