Skip to Content
Merck
All Photos(1)

Documents

1.04190

Sigma-Aldrich

Fuchsin acid (C.I. 42685)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H17N3Na2O9S3
CAS Number:
Molecular Weight:
585.54
UNSPSC Code:
12352106
NACRES:
NA.21

form

solid

Quality Level

composition

Dye, ≥60% spectrophotometric assay

pH

3-4 (20 °C, 10 g/L in H2O)

mp

>130 °C (Decomposition)

solubility

10 g/L

bulk density

920 kg/m3

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

2-30°C

InChI

1S/C20H19N3O9S3.2Na/c1-10-6-13(9-18(20(10)23)35(30,31)32)19(11-2-4-14(21)16(7-11)33(24,25)26)12-3-5-15(22)17(8-12)34(27,28)29;;/h2-9,23H,21-22H2,1H3,(H,24,25,26)(H,27,28,29)(H,30,31,32);;/q;2*+1/p-2

InChI key

WZRZTHMJPHPAMU-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Fuchsin acid (C.I. 42685) for microscopy Certistain®, is a triarylmethane dye used, among others, in trichrome connective tissue stainings acc. to Mallory and van Gieson. Mallory developed a staining procedure for visualization of collagenous connective tissue, which has been modified and improved over the years.It enables a good differentiation of individual tissue components, provided that the tissue has been fixed in Zenker’s solution. As CERTISTAIN® dye it is chemically quality-certified according to highest standards and strict specifications.
for microscopy Certistain®

Analysis Note

Dye content (spectrophotometrically): ≥ 60 %
Identity (UV/VIS-Spectrum): passes test
Absorption maximum λ max (Hydrochloric acid 0.0025 mol/l): 540 - 549 nm
Spec. Absorptivity A 1%/1cm (λmax; 0.005 g/l; hydrochloric acid 0.0025 mol/l): 780 - 1310
TLC-Test: passes test
Loss on drying (110 °C): ≤ 10 %
Suitability for microscopy: passes test

Legal Information

CERTISTAIN is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service