Skip to Content
Merck
All Photos(1)

Key Documents

1.03935

Supelco

Iron(III) phosphate

for analysis calcined (max. 0.001% SO₄) EMSURE®

Synonym(s):

Iron(III) phosphate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
FeO4P
CAS Number:
Molecular Weight:
150.82
UNSPSC Code:
12352302
EC Index Number:
233-149-7
form:
solid

Quality Level

product line

EMSURE®

form

solid

composition

Fe, ≥33% iodometric
PO4, ≤68% gravimetric

loss

≤0.1% loss on ignition, 700 °C

density

2.87 g/cm3 at 20 °C

bulk density

900 kg/m3

anion traces

sulfur compounds (as SO42-): ≤0.001%

storage temp.

2-30°C

InChI

1S/Fe.H3O4P/c;1-5(2,3)4/h;(H3,1,2,3,4)/q+3;/p-3

InChI key

WBJZTOZJJYAKHQ-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Analysis Note

Assay (iodometric, Fe): ≥ 33 %
Assay (gravimetric, PO₄): ≤ 68 %
Total sulfur (as SO₄): ≤ 0.001 %
Loss on ignition (700 °C): ≤ 0.1 %

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service