Skip to Content
Merck
All Photos(1)

Documents

1.02754

Millipore

GranuCult® Prime Vegetable Peptone Broth Halal

Suitable for use in Halal and Kosher certified processes, JP 5.02

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106213
NACRES:
NA.74

Agency

EP 2.6.13
JP 5.02
USP 62

Quality Level

sterility

irradiated

form

granular

feature

granulated

manufacturer/tradename

GranuCult® prime

technique(s)

microbiological culture: suitable

pH

7.3 ± 0.2 (25.0 °C)

application(s)

aseptic process simulation
pharmaceutical

compatibility

stand alone

storage temp.

15-25°C

suitability

nonselective for

General description

GranuCult® Prime Vegetable Peptone Broth Halal is a non-animal origin alternative to Tryptic Soy Broth (TSB). The media is granulated to ensure excellent solubility in cold water. The product is gamma irradiated and triple bagged to allow transfer to controlled areas.
Peptones provide nitrogen, vitamins, and minerals for the growing microflora. The natural sugars, derived from the peptones promote bacterial growth, and sodium chloride maintains the osmotic balance of the medium.

Application

GranuCult® Prime Vegetable Peptone Broth is suitable for aseptic process simulation, also known as Media fill test.
The media is used to cultivate and isolate fastidious or non-fastidious microorganisms from test samples and for the growth of both aerobic and anaerobic microflora.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Halal-certified culture media prevent non-halal ingredient contamination, enabling microbiological QC testing without losing certification.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service