1.01398
Malachite green oxalate (C.I.42000)
for microscopy and for microbiology
Synonym(s):
Malachite green oxalate (C.I.42000)
About This Item
Recommended Products
Quality Level
form
solid
composition
Dye, ≥90% spectrophotometric assay
pH
2.4 (24 °C, 10 g/L in H2O)
mp
159 °C
solubility
110 g/L
bulk density
400‑500 kg/m3
εmax
1730-1960 at 616-620 nm in water at 0.003 g/L
application(s)
diagnostic assay manufacturing
hematology
histology
storage temp.
2-30°C
InChI
1S/2C23H25N2.3C2H2O4/c2*1-24(2)21-14-10-19(11-15-21)23(18-8-6-5-7-9-18)20-12-16-22(17-13-20)25(3)4;3*3-1(4)2(5)6/h2*5-17H,1-4H3;3*(H,3,4)(H,5,6)/q2*+1;;;/p-2
InChI key
CNYGFPPAGUCRIC-UHFFFAOYSA-L
General description
Analysis Note
Identity
- oxalate:
passes test
- UV/VIS-Spectrum: passes test
Absorption maximum λ max (water): 616 - 620 nm
Spec. Absorptivity A 1%/1cm (λmax; 0.003 g/l; water): 1730 - 1960
TLC-Test: passes test
Loss on drying (50 °C / <1 hPa): ≤ 7%
Suitability for microbiology: passes test
Suitability for microscopy: passes test
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Repr. 2
Storage Class Code
6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service