1.01368
Brilliant cresyl blue zinc chloride double salt
for microscopy Certistain®
Synonym(s):
Brilliant cresyl blue zinc chloride double salt
About This Item
Recommended Products
Quality Level
form
solid
composition
Dye, ≥60% spectrophotometric assay
pH
5.5 (20 °C, 5 g/L in H2O)
mp
195-198 °C
solubility
30 g/L
bulk density
560 kg/m3
εmax
≥1500 at 623-628 nm in 50% ethanol at 0.005 g/L
application(s)
diagnostic assay manufacturing
hematology
histology
storage temp.
2-30°C
InChI
1S/2C17H20N3O.4ClH.Zn/c2*1-4-20(5-2)12-6-7-14-16(9-12)21-17-10-13(18)11(3)8-15(17)19-14;;;;;/h2*6-10H,4-5,18H2,1-3H3;4*1H;/q2*+1;;;;;+2/p-4
InChI key
SIHASHFMDFXGSX-UHFFFAOYSA-J
Looking for similar products? Visit Product Comparison Guide
General description
As an IVD product and CE registered, it can be used hassle-free in diagnostics and laboratory accreditations. For more details, please see instructions for use (IFU). The IFU can be downloaded from this webpage.
Analysis Note
Identity
- UV/VIS-Spectrum:
passes test
- Zinc: passes test
Absorption maximum λ max (ethanol 50 %): 623 - 628 nm
Spec. Absorptivity A 1%/1cm (λmax; 0.005 g/l; ethanol 50 %): ≥ 1500
TLC-Test: passes test
Loss on drying (110 °C): ≤ 5 %
Suitability for microscopy: passes test
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service