05-806-AF555
Anti-phospho-Histone H3 (Ser10) Antibody, clone 3H10, Alexa Fluor™ 555
clone 3H10, from mouse, ALEXA FLUOR™ 555
Synonym(s):
H3S10P, Histone H3 (phospho Ser10), H3 histone, family 3A, H3.3A, H3 histone, family 3B, H3.3B
About This Item
Recommended Products
biological source
mouse
Quality Level
conjugate
ALEXA FLUOR™ 555
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
3H10, monoclonal
species reactivity
human
technique(s)
immunocytochemistry: suitable
isotype
IgG1κ
NCBI accession no.
UniProt accession no.
shipped in
wet ice
target post-translational modification
phosphorylation (pSer10)
Gene Information
human ... H3C1(8350)
General description
Specificity
Immunogen
Application
The unconjugated antibody (Cat. No. 05-806) is shown to be suitable also for peptide inhibition assay (PIA), flow cytometry, immunocytochemistry, immunofluorescence, multiplex assay, Western blotting applications.
Quality
Immunocytochemistry Analysis: A 1:100 dilution of this antibody detected phospho-Histone H3 (Ser10) in HeLa cells.
Target description
Physical form
Other Notes
Legal Information
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service