Skip to Content
Merck
All Photos(1)

Key Documents

KEY090496932

Key Organics / BIONET

O-[(3-chlorophenyl)methyl]hydroxylamine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
194.06
MDL number:

mp

206 - 212

Iupac Name

1-[(aminooxy)methyl]-3-chlorobenzene hydrochloride

Inchi Code

1S/C7H8ClNO.ClH/c8-7-3-1-2-6(4-7)5-10-9;/h1-4H,5,9H2;1H

InChI key

PZPCEIVCWGWHCC-UHFFFAOYSA-N

Purity

95%

Country of Origin

GB

Physical Form

Solid

Product Link

https://www.keyorganics.net/sku/10F-930

Melting Point

206 - 212

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Codes

Precautionary statements

MSDS

https://www.keyorganics.net/sds/10F-930


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library