Skip to Content
Merck
All Photos(1)

Key Documents

ENA580351371

Enamine

7-methoxyquinoline-8-sulfonyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
257.7
MDL number:
Pricing and availability is not currently available.

Iupac Name

7-methoxyquinoline-8-sulfonyl chloride

Inchi Code

1S/C10H8ClNO3S/c1-15-8-5-4-7-3-2-6-12-9(7)10(8)16(11,13)14/h2-6H,1H3

InChI key

BOYGXUWPYKJSSH-UHFFFAOYSA-N

Purity

95

Country of Origin

UA

Product Link

https://www.enaminestore.com/catalog/EN300-319583

MSDS

https://www.enaminestore.com/catalog/EN300-319583


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library