Skip to Content
Merck
All Photos(1)

Key Documents

ENA375227824

Enamine

6-phenoxy-2-propyl-1,4-dihydroquinolin-4-one

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
279.34
MDL number:

Iupac Name

6-phenoxy-2-propyl-4-quinolinol

Inchi Code

1S/C18H17NO2/c1-2-6-13-11-18(20)16-12-15(9-10-17(16)19-13)21-14-7-4-3-5-8-14/h3-5,7-12H,2,6H2,1H3,(H,19,20)

InChI key

ZSNFTBBLVPRSRI-UHFFFAOYSA-N

storage temp.

RT

Purity

95

Country of Origin

UA

Physical Form

powder

Product Link

https://www.enaminestore.com/catalog/EN300-115327

MSDS

https://www.enaminestore.com/catalog/EN300-115327


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library