Skip to Content
Merck
All Photos(1)

Documents

ENA024296142

Enamine

1-[(5-sulfanyl-1,3,4-oxadiazol-2-yl)methyl]azepan-2-one

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
227.29
MDL number:

Iupac Name

1-[(5-sulfanyl-1,3,4-oxadiazol-2-yl)methyl]-2-azepanone

Inchi Code

1S/C9H13N3O2S/c13-8-4-2-1-3-5-12(8)6-7-10-11-9(15)14-7/h1-6H2,(H,11,15)

InChI key

FRNUPZXKVBWLIM-UHFFFAOYSA-N

Purity

95

Country of Origin

UA

Product Link

https://www.enaminestore.com/catalog/EN300-10106

MSDS

https://www.enaminestore.com/catalog/EN300-10106


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service