Skip to Content
Merck
All Photos(1)

Key Documents

CH6371383664

Chem Impex International, Inc.

(2-Methyl-imidazo[1,2-a]pyridin-3-yl)methylamine

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
161.21
MDL number:

mp

61-67 C

Iupac Name

(2-methylimidazo[1,2-a]pyridin-3-yl)methanamine

Inchi Code

1S/C9H11N3/c1-7-8(6-10)12-5-3-2-4-9(12)11-7/h2-5H,6,10H2,1H3

InChI key

XMHJAFWCJJRDBR-UHFFFAOYSA-N

storage temp.

Store at 0-8 C

Purity

96%

Shipping Temperature

RT

Physical Form

Light yellow crystals

Product Link

https://www.chemimpex.com/product/productinfo/2-methyl-imidazo-1-2-a-pyridin-3-yl-methylamine/10386?cid=0

Melting Point

61-67 C

MSDS

https://www.chemimpex.com/product/productinfo/2-methyl-imidazo-1-2-a-pyridin-3-yl-methylamine/10386?cid=0


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library