CH4424845927
ChemBridge Corporation
1-acetyl-1,4-diazepane dihydrochloride hydrate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Molecular Weight:
233.14
MDL number:
Iupac Name
1-(1,4-diazepan-1-yl)ethan-1-one dihydrochloride hydrate
Inchi Code
1S/C7H14N2O.2ClH.H2O/c1-7(10)9-5-2-3-8-4-6-9;;;/h8H,2-6H2,1H3;2*1H;1H2
InChI key
QIPPADZXMLWZGU-UHFFFAOYSA-N
storage temp.
RT
Salt Data
2HCl H2O
Purity
95%
Country of Origin
KZ
Shipping Temperature
RT
Physical Form
solid
Product Link
https://www.hit2lead.com/building-blocks/4028076
MSDS
https://www.hit2lead.com/sds/4028076_sds.pdf
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.