Skip to Content
Merck
All Photos(2)

Documents

857466W

Avanti

AEA-d11

Avanti Research - A Croda Brand 857466W, methanol solution

Synonym(s):

C20:4 anandamide-d11

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H26D11NO2
CAS Number:
Molecular Weight:
358.60
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

methanol solution

packaging

pkg of 1 × 1 mL (857466W-500ug)
pkg of 2 × 1 mL (857466W-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 857466W

concentration

0.5 mg/mL (857466W-1mg)
0.5 mg/mL (857466W-500ug)

shipped in

dry ice

storage temp.

−20°C

SMILES string

O=C(CCC/C=C\C/C=C\C/C=C\C/C=C\C([2H])(C([2H])(C([2H])(C([2H])(C([2H])([2H])[2H])[2H])[2H])[2H])[2H])NCCO

General description

Anandamide (AEA) is an agonist of the cannabinoid and non-cannabinoid receptors. It is an N-acyl ethanolamide derivative of C20:4 fatty acid, which is also known as arachidonic acid. AEA is a lipid neurotransmitter synthesized and released by the brain nerve cells as well as macrophages.

Biochem/physiol Actions

Anandamide (AEA) is involved in many physiological and pathophysiological functions, such as regulation of energy consumption and balance. It is also involved in initiation and control of inflammation and modulatory effects on sensory and autonomic nerve signaling.

Packaging

2 mL Amber Glass Sealed Ampule (857466W-1mg)
2 mL Amber Glass Sealed Ampule (857466W-500ug)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Storage Class Code

3 - Flammable liquids

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anandamide and its metabolites: what are their roles in the kidney?
Ritter J K, et al.
Frontiers in Bioscience, 8, 264-264 (2016)
A biosynthetic pathway for anandamide
Liu J, et al.
Proceedings of the National Academy of Sciences of the USA, 103(36), 13345-13350 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service