Skip to Content
Merck
All Photos(2)

Documents

810178P

Avanti

06:0-12:0 NBD PA

Avanti Research - A Croda Brand 810178P, powder

Synonym(s):

1-hexanoyl-2-{12-[(7-nitro-2-1,3-benzoxadiazol-4-yl)amino]dodecanoyl}-sn-glycero-3-phosphate (ammonium salt)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H46N5O11P
CAS Number:
Molecular Weight:
647.65
UNSPSC Code:
12352211

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (810178P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 810178P

shipped in

dry ice

storage temp.

−20°C

Packaging

5 mL Clear Glass Sealed Ampule (810178P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service