Skip to Content
Merck
All Photos(2)

Documents

700075P

Avanti

8(9)-dehydrocholesterol

Avanti Research - A Croda Brand

Synonym(s):

cholesta-5,8(9)-dien-3β-ol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H44O
CAS Number:
Molecular Weight:
384.64
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (700075P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand

shipped in

dry ice

storage temp.

−70°C

SMILES string

C[C@]1(CC2)C(CCC1C(CCCC(C)C)C)C3=C2[C@](CC[C@H](O)C4)(C)C4=CC3

InChI

1S/C27H44O/c1-18(2)7-6-8-19(3)23-11-12-24-22-10-9-20-17-21(28)13-15-26(20,4)25(22)14-16-27(23,24)5/h9,18-19,21,23-24,28H,6-8,10-17H2,1-5H3/t19-,21+,23-,24+,26+,27-/m1/s1

InChI key

VUKORTMHZDZZFR-BXAZICILSA-N

General description

8(9)-dehydrocholesterol is an isomer of 7-dehydrocholesterol and non-polar oxysterol metabolite. It is oxidized to Δ8 analog by mitochondrial enzymes.

Application

8(9)-dehydrocholesterol has been used as a component of sterol standard in liquid chromatography–mass spectrometry (LC-MS) for quantification of lipids from cultured cells.

Biochem/physiol Actions

8(9)-dehydrocholesterol (8-DHC) elevated levels are toxic to cells and is implicated in Smith-Lemli-Opitz syndrome (SLOS). 8-DHC profiling helps in understanding SLOS phenotype.

Packaging

5 mL Amber Glass Screw Cap Vial (700075P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

William J Griffiths et al.
The Journal of steroid biochemistry and molecular biology, 169, 77-87 (2016-03-16)
Smith-Lemli-Opitz syndrome (SLOS) is a severe autosomal recessive disorder resulting from defects in the cholesterol synthesising enzyme 7-dehydrocholesterol reductase (Δ7-sterol reductase, DHCR7, EC 1.3.1.21) leading to a build-up of the cholesterol precursor 7-dehydrocholesterol (7-DHC) in tissues and blood plasma. Although
Josi Herron et al.
Current protocols in toxicology, 78(1), e65-e65 (2018-10-16)
Cholesterol and cholesterol-derived oxysterols are critical for embryonic development, synapse formation and function, and myelination, among other biological functions. Indeed, alterations in levels of cholesterol, sterol precursors, and oxysterols result in a variety of developmental disorders, emphasizing the importance of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service