Skip to Content
Merck
All Photos(1)

Key Documents

ATC311765434

Atlantic Research Chemicals Ltd

Benzoin hemisuccinate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
298.34
MDL number:
Pricing and availability is not currently available.

Iupac Name

4-(2-oxo-1,2-diphenylethoxy)butanoic acid

Inchi Code

1S/C18H18O4/c19-16(20)12-7-13-22-18(15-10-5-2-6-11-15)17(21)14-8-3-1-4-9-14/h1-6,8-11,18H,7,12-13H2,(H,19,20)

InChI key

XGODCRXAWGHJKX-UHFFFAOYSA-N

Purity

95

Country of Origin

GB

Product Link

https://atlantic-chemicals.com/product/?code=CA10132

MSDS

https://atlantic-chemicals.com/product/?code=CA10132


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library