Skip to Content
Merck
All Photos(1)

Key Documents

APOH11A9D1C3

Apollo Scientific Ltd

Ethyl 5-phenyl-1,3-oxazole-4-carboxylate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
217.22
MDL number:

Iupac Name

ethyl 5-phenyl-1,3-oxazole-4-carboxylate

Inchi Code

1S/C12H11NO3/c1-2-15-12(14)10-11(16-8-13-10)9-6-4-3-5-7-9/h3-8H,2H2,1H3

InChI key

LFBYYERWTHYROG-UHFFFAOYSA-N

storage temp.

Ambient Temperature

Country of Origin

GB

Shipping Temperature

Ambient Temperature

Physical Form

Solid

Product Link

https://store.apolloscientific.co.uk/product/ethyl-5-phenyl-13-oxazole-4-carboxylate

Pictograms

Signal Word

TBC

Hazard Statements

Precautionary statements

MSDS

https://store.apolloscientific.co.uk/product/ethyl-5-phenyl-13-oxazole-4-carboxylate


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library