Skip to Content
Merck
All Photos(1)

Key Documents

APO455832824

Apollo Scientific Ltd

4'-(Benzyloxy)-[1,1'-biphenyl]-2-amine

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
275.35
MDL number:

bp

116-118

Iupac Name

4'-(benzyloxy)[1,1'-biphenyl]-2-amine

Inchi Code

1S/C19H17NO/c20-19-9-5-4-8-18(19)16-10-12-17(13-11-16)21-14-15-6-2-1-3-7-15/h1-13H,14,20H2

InChI key

XKIPQWRPJDHCCW-UHFFFAOYSA-N

storage temp.

Ambient Temperature

Purity

98%

Country of Origin

GB

Shipping Temperature

Ambient Temperature

Physical Form

Solid

Product Link

https://store.apolloscientific.co.uk/product/4-benzyloxy-11-biphenyl-2-amine

Boiling point

116-118

Pictograms

Exclamation mark

Signal Word

Warning

Precautionary statements

MSDS

https://store.apolloscientific.co.uk/product/4-benzyloxy-11-biphenyl-2-amine


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library