Skip to Content
Merck
All Photos(1)

Key Documents

AMBH97BA012E

Ambeed, Inc.

6-Chloro-5-(trifluoromethyl)pyridine-3-sulfonyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
280.05
MDL number:
Pricing and availability is not currently available.

Iupac Name

6-chloro-5-(trifluoromethyl)-3-pyridinesulfonyl chloride

Inchi Code

1S/C6H2Cl2F3NO2S/c7-5-4(6(9,10)11)1-3(2-12-5)15(8,13)14/h1-2H

InChI key

UUVXRPZDCPBGQS-UHFFFAOYSA-N

storage temp.

Inert atmosphere,Store in freezer, under -20C

Purity

98%

Country of Origin

CN

Shipping Temperature

Cold-chain transportation

Physical Form

solid

Product Link

https://www.ambeed.com/products/928324-59-8.html

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary statements

MSDS

https://www.ambeed.com/products/928324-59-8.html


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library