AMBH2D6F8B89
Ambeed, Inc.
5-(Trifluoromethyl)picolinaldehyde
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
5-(trifluoromethyl)-2-pyridinecarbaldehyde
Inchi Code
1S/C7H4F3NO/c8-7(9,10)5-1-2-6(4-12)11-3-5/h1-4H
InChI key
JJDDVGAESNBKMY-UHFFFAOYSA-N
storage temp.
Inert atmosphere,2-8C
Purity
95%
Country of Origin
CN
Shipping Temperature
Normal
Physical Form
Colorless to yellow to brown Liquid or Semi-Solid or Solid or Lump
Product Link
https://www.ambeed.com/products/31224-82-5.html
Signal Word
Warning
Hazard Statements
Precautionary statements
MSDS
https://www.ambeed.com/products/31224-82-5.html
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.