Z760331
Ohaus® MB-23 and MB-25 moisture analyzers
Model MB25, AC/DC input 230 V AC, European 2-pin plug
Synonym(s):
moisture meters
All Photos(1)
About This Item
Recommended Products
manufacturer/tradename
Ohaus® 80252472 EU
weighing capacity measuring range
110 g
parameter
50-160 °C temp. limit
W × H × depth
17 cm × 13 cm × 28 cm
platform diam.
90 mm
General description
The MB25 has a halogen lamp for fast, even heating.
The Moisture analyzers combine high quality construction and easy setup. The LCD display is backlit, and it has a standard RS232 port.
- easy to clean design
- heating chamber is low maintenance
- compact footprint
Legal Information
Ohaus is a registered trademark of Ohaus Corp.