Documents
Z756989
Sigma-Aldrich
Wrap-it ties
Tear away version, blue Nylon, L 6 in.
Synonym(s):
tie wrap, wrap tie
About This Item
UNSPSC Code:
41100000
NACRES:
NB.35
material
blue Nylon
L
6 in.
General description
Self-locking, one-piece nylon construction. Excellent for securing rubber septa to bottles and flask joints, attachment of tubing to hose barbs or glassware, and for closing plastic bags or drum liners. Use with Wrap-it tie installing tool.
Tear-off version is blue in color and is easily removed from septum with a quick pulling motion.