Z681938
CellCrown™ inserts
24 well plate inserts with 1.2 μm nylon filter, non-sterile
Synonym(s):
3D, 3D Cell Culture, 3D scaffolds, cell scaffolds
About This Item
sterility
non-sterile
feature
lid
manufacturer/tradename
Scaffdex C12001F
size
24 wells
color
none
binding type
non-treated surface
Looking for similar products? Visit Product Comparison Guide
General description
Sterilization Conditions:
Temperature.......121 °C / 250 °F
Time..................15 min.
Drying time.........15 min.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service