Z678627
2mag MIX 6 multi position stirrer
AC/DC input 100 / 240 V AC, Schuko plug
Synonym(s):
induction multi-position stirrer, magnetic induction stir plate, magnetic wear-free stirrer, multi-position stirrer
About This Item
Recommended Products
material
stainless steel plate
feature
CE compliant
positions 6
packaging
pack of 1 ea
manufacturer/tradename
2mag 30006
parameter
100-2000 rpm speed
plate size × W × L
× 245 mm × 400 mm
stirrer volume
1-3000 mL , per stirring position
General description
Powerful 6 stirring positions, 100% maintenance-free and wear-free by inductive 2mag-Magnetic-Drive concept, extremely wide speed range of 100 to 2,000 rpm, 100% synchronized speed, jerk-free stirring even at low speeds, 4-step power settings of the stirrer, high power setting for viscous liquids and reduced power setting for long-term operation without any heating effects caused by the stirrer.