Skip to Content
Merck
All Photos(1)

Documents

Z677108

Millipore

Amicon Ultra 0.5 mL centrifugal filters

MWCO 10 kDa

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103900
NACRES:
NB.24

material

polypropylene tube
regenerated cellulose membrane
styrene-butadiene housing

sterility

non-sterile

manufacturer/tradename

Millipore UFC501096

tube size

1.5 mL

working volume

1.5 mL

pore size

10 kDa MWCO

Looking for similar products? Visit Product Comparison Guide

General description

With their vertical membrane design, Amicon Ultra 0.5 mL filters deliver great performance and lower spin times.

  • Concentrates 500 μL down to 15 μL
  • Concentration factor 25 x to 30 x
  • High sample recovery (greater than 90% of dilute starting solution)
  • Fast processing time - 10 to 30 minutes
  • Consistent recoveries thanks to the reverse spin capability
  • Reverse spin capability provides consistent recoveries

The Amicon Ultra-0.5 device is supplied with two microcentrifuge tubes. During operation, one tube is used to collect filtrate; the other tube is to recover the concentrated sample.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

  1. Which document(s) contains shelf-life or expiration date information for a given product?

    If available for a given product, the recommended re-test date or the expiration date can be found on the Certificate of Analysis.

  2. How do I get lot-specific information or a Certificate of Analysis?

    The lot specific COA document can be found by entering the lot number above under the "Documents" section.

  3. How do I find price and availability?

    There are several ways to find pricing and availability for our products. Once you log onto our website, you will find the price and availability displayed on the product detail page. You can contact any of our Customer Sales and Service offices to receive a quote.  USA customers:  1-800-325-3010 or view local office numbers.

  4. What is the Department of Transportation shipping information for this product?

    Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product. 

  5. How do I use the Amicon Ultra 0.5ML Centrifugal Filters?

    The vendor instruction guide for the Amicon Ultra 0.5ML Centrifugal Filters is attached.

  6. My question is not addressed here, how can I contact Technical Service for assistance?

    Ask a Scientist here.

Rachel C Childers et al.
American journal of physiology. Heart and circulatory physiology, 316(3), H596-H608 (2018-12-24)
Hemodynamic load regulates cardiac remodeling. In contrast to pressure overload (increased afterload), hearts subjected to volume overload (VO; preload) undergo a distinct pattern of eccentric remodeling, chamber dilation, and decreased extracellular matrix content. Critical profibrotic roles of cardiac fibroblasts (CFs)
Jules Petit et al.
Journal of immunology (Baltimore, Md. : 1950), 203(1), 216-224 (2019-05-28)
Trained immunity is a form of innate immune memory best described in mice and humans. Clear evidence of the evolutionary conservation of trained immunity in teleost fish is lacking. Given the evolutionary position of teleosts as early vertebrates with a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service