Key Documents
Z566020
Sigma-Aldrich
Pure-Pac® II dispense coupler kit
3/8in. tube end
Select a Size
1 KIT
£1,110.00
£1,110.00
Please contact Customer Service for Availability
Select a Size
Change View
1 KIT
£1,110.00
About This Item
UNSPSC Code:
41100000
NACRES:
NB.01
£1,110.00
Please contact Customer Service for Availability
General description
The Sigma-Aldrich Pure-Pac II returnable containers require the Micromatic MacroValve coupler for dispensing chemical. The Dispense Head Coupler Kit includes the MacroValve coupler along with the standard components needed to connect to either 1/2in. or 3/8in. diameter transfer lines. All of the hardware in the kit is supplied in a foam-lined plastic case. See Technical Bulletin AL-236 for assembly instructions.
Legal Information
Pure-Pac is a registered trademark of Merck KGaA, Darmstadt, Germany
Kit Components Also Available Separately
Product No.
Description
SDS
  • Z104388PTFE sealing tape, W 1/2 in.SDS
  • Male connector with double ferrules and nut, SS, ferrule size 3/8 in., NPTM 1/2 in.
  • Z560723Micromatic MacroValve coupler for returnable containersSDS
  • Swagelok® hex reducing coupling, pkg of 1 ea
  • Swagelok® street elbow, pkg of 1 ea
  • Z565547Stainless steel hose connector, 1/4in. male NPT, 1/4in. hose I.D., pkg of 1 eaSDS
Choose from one of the most recent versions:
Certificates of Analysis (COA)