Z565202
Büchi® Rotavapor® R-210 evaporator with jack and water bath, 29/32 joint, 240V
Advanced, condenser, cold-trap reflux, plastic-coated glass
All Photos(1)
About This Item
UNSPSC Code:
41100000
material
plastic-coated glass
feature
CE compliant
CSA approved
condenser cold-trap reflux
manufacturer/tradename
Büchi 23011R110
General description
R-210 Features:
1. Four different glassware assemblies
Supplied with a standard 29/32 joint on the evaporator flask and a 35/20 ball joint on the receiving flask.
a. Diagonal condenser assembly- For simple distillation of solvents and general use. Tap-water-cooled. Inlet feed tube for continuous feed of volumes exceeding flask capacity.
b. Vertical condenser assembly- For standard distillations and for high-boiling solvents (>100°C). Tap-water-cooled. Inlet feed tube for continuous feed of volumes exceeding flask capacity.
c. Cold-trap condenser assembly- Two-piece cold-trap condenser style is ideal for low-boiling solvents. Uses mechanical refrigeration or dry ice and acetone.
d. Cold-trap reflux condenser assembly- Two-piece cold-trap condenser plus reflux distribution head for use with low-boiling solvents with reflux.
2. Three different Vacuum Controller options
a. Basic- no vacuum controller
b. Advanced- comes with V-850 vacuum controller. See Z616850 listing for specifications.
c. Professional- comes with V-855 vacuum controller. See Z616877 listing for specifications.
3. Two different Glassware options- Glass or Plastic-coated Glass
- PTFE composite vacuum seal for long service life and effective sealing
- Space-saving modular design lets you add integrated vacuum controllers and run preset time/pressure gradients
- Motorized, quick-action servo-jack raises and lowers glassware effortlessly
- Wide-range, PTFE-coated 4L heating bath, model B-491, with digital temperature display, uses oil or water, 20 to 180°C (68 to 356°F)
- Enhanced safety: insulated heating bath, Plastic-coated glassware, and optional remote control and safety shield
- Process samples from 50 mL to 3 L
1. Four different glassware assemblies
Supplied with a standard 29/32 joint on the evaporator flask and a 35/20 ball joint on the receiving flask.
a. Diagonal condenser assembly- For simple distillation of solvents and general use. Tap-water-cooled. Inlet feed tube for continuous feed of volumes exceeding flask capacity.
b. Vertical condenser assembly- For standard distillations and for high-boiling solvents (>100°C). Tap-water-cooled. Inlet feed tube for continuous feed of volumes exceeding flask capacity.
c. Cold-trap condenser assembly- Two-piece cold-trap condenser style is ideal for low-boiling solvents. Uses mechanical refrigeration or dry ice and acetone.
d. Cold-trap reflux condenser assembly- Two-piece cold-trap condenser plus reflux distribution head for use with low-boiling solvents with reflux.
2. Three different Vacuum Controller options
a. Basic- no vacuum controller
b. Advanced- comes with V-850 vacuum controller. See Z616850 listing for specifications.
c. Professional- comes with V-855 vacuum controller. See Z616877 listing for specifications.
3. Two different Glassware options- Glass or Plastic-coated Glass
Linkage
View the Selection Guide of available Rotavapors.
Legal Information
Büchi is a registered trademark of Büchi Labortechnik AG
Rotavapor is a registered trademark of Büchi Labortechnik AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)