Documents
Z562521
LabJaws end-to-end lattice rod connector
jaw capacity 0-13 mm , 0-51 inches, aluminum
About This Item
UNSPSC Code:
41100000
NACRES:
NB.35
material
aluminum
manufacturer/tradename
Troemner 916133
jaw capacity
0-13 mm , 0-51 inches
General description
Corrosion resistant. Increases the length of lab frame rods and clamps with perfect alignment. Comes with 2 set screws.
Legal Information
LabJaws is a trademark of Henry Troemner LLC