Key Documents
Z543853
Sigma-Aldrich
Compression tubing fitting for VerSA-Flow dispenser
tubing O.D. 1/8 in.
About This Item
UNSPSC Code:
41105108
tubing O.D.
1/8 in.
General description
Adapts rigid wall tubing of various sizes to VerSA-Flow caps. Fitting includes nut with ferrules and PBT bushing with 1/4 in. NPTM threads.
Legal Information
VerSA-Flow is a trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)