Z415499
Water-cooled distilling head
joint: ST/NS 24/40, Overall H 400 mm, O.D. 54 mm
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Joint
joint: ST/NS24/40
O.D.
54 mm
Overall H
400 mm
side arm clearance
225 mm
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Useful for removal of solvents from reaction mixtures or concentrations from solutions prior to crystallization.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service