Documents
Z279978
Sigma-Aldrich
Norprene® A-60-G laboratory tubing
I.D. × O.D. 5/16 in. × 7/16 in.
Synonym(s):
Saint Gobain AFL00022, Tygon A-60-G
About This Item
UNSPSC Code:
41105108
NACRES:
NB.35
packaging
pkg of 50 feet
manufacturer/tradename
Tygon
I.D. × O.D.
5/16 in. × 7/16 in.
wall size
1/16 in.
General description
This chemical pump tubing is a thermoplastic elastomer tubing with chemical compatibilities approaching fluoroelastomers and silicone tubing. Black, rubber-like tubing is heat sealable. Max. temp.: 275°F (135°C).
  • Chemical resistance to oxidizing agents, acids, and alkalies
  • Resistance to flex-fatigue, will not weaken or crack
Legal Information
Norprene is a registered trademark of Saint-Gobain Abrasives, Inc.