Key Documents
Z154245
Compressor-type tubing clamp
heavy-duty, nickel-plated brass
About This Item
UNSPSC Code:
41105108
NACRES:
NB.35
opening W × H
35 mm × 30 mm
General description
  • Heavy-duty, nickel-plated brass
  • Opening: 35 W × 30 H (mm)
  • Lower bar held in place by threaded cap; may be swung out to insert tubing
  • 6 mm screw with 35 mm knurled finger grip
Choose from one of the most recent versions:
Certificates of Analysis (COA)