Skip to Content
Merck
All Photos(1)

Documents

WHA2651500

Whatman® indicator papers

Starch iodide test paper book, book of 20 strips

Synonym(s):

Z753351

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41113034
NACRES:
NB.79

reaction suitability

reagent type: oxidant

manufacturer/tradename

Whatman 2651-500
Whatman Article No. 28418935 (US reference)

packaging

pkg of 10 books of 20 strips

detection method

colorimetric

Looking for similar products? Visit Product Comparison Guide

General description

pH indicator and test papers to meet specific needs, offering quality, convenience and ease-of-use. We offer a range of pH indicator and test papers to meet specific needs. These high-quality papers combine ease-of-use with accuracy and consistency.

Lead acetate test paper

Used for detecting hydrogen sulfide, this rapid qualitative test paper, when wetted with distilled water, can detect as little as 5 ppm of H2S in the atmosphere or in a gas stream. Hydrogen peroxide can be detected with this paper by preblackening the paper in H2S. Concentrations as low as 4 ppm can be detected.

Potassium iodide test paper

Used for detecting chlorine and other oxidizing agents. In acid solution, oxidizing agents react with the iodide in the test paper to liberate iodine. The paper will turn blue in the presence of an oxidizing agent (e.g. CI2, Br2, H2O2, HNO2 etc.).

Universal indicator papers

Universal indicator papers have been impregnated with a mixture of several indicators. On contact with the sample solution they assume a particular color. A check against the color comparison table supplied allows the pH to be determined.

Features and Benefits

  • Instant pH readings.
  • Accurate for a wide range of routine pH testing.
  • Inexpensive.
  • Convenient and portable for field use.

Other Notes

Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.

Legal Information

Whatman is a registered trademark of Cytiva

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service