About This Item
biological source
synthetic
grade
Kosher
Assay
≥98%
refractive index
n20/D 1.502 (lit.)
bp
262 °C (lit.)
density
1.08 g/mL at 25 °C (lit.)
application(s)
flavors and fragrances
Organoleptic
lavender
SMILES string
CCC(=O)OCCOc1ccccc1
InChI
1S/C11H14O3/c1-2-11(12)14-9-8-13-10-6-4-3-5-7-10/h3-7H,2,8-9H2,1H3
InChI key
FKTSMIXZGPUSJB-UHFFFAOYSA-N
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Flash Point(F)
219.2 °F - closed cup
Flash Point(C)
104 °C - closed cup
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service